Learn More
Waters Corp Waters Sep-Pak Plus Light Cartridge, tC18, 145 mg Sorbent, 37-55 μm
Catalog No. 50785398 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These Sep-Pak tC18 Plus Light Cartridges are engineered for solid phase extraction, featuring a silica-based, trifunctional bonded phase with strong hydrophobicity. They provide enhanced hydrolytic stability, suitable for sample preparation in HPLC, GC, and LC-MS. The cartridges reduce steps, streamline protocols, and simplify handling, aiding in emulsion challenges and improving safety. They are constructed with a high-purity polypropylene syringe barrel body, compatible with multi-position vacuum manifolds, and comply with ISO 9001 standards.
  • Features packing material held by two polyethylene frits.
  • Designed with a female Luer inlet that accepts a male Luer tip.
  • Molded for precise dimensions, suitable for automation equipment.
  • Sorbent chemistries tested for chromatographic activities, retention, and selectivity.
Catalog No. 50-785-398 Supplier Waters Corp Supplier No. WAT036805
These Sep-Pak tC18 Plus Light Cartridges are engineered for solid phase extraction, featuring a silica-based, trifunctional bonded phase with strong hydrophobicity. They provide enhanced hydrolytic stability, suitable for sample preparation in HPLC, GC, and LC-MS. The cartridges reduce steps, streamline protocols, and simplify handling, aiding in emulsion challenges and improving safety. They are constructed with a high-purity polypropylene syringe barrel body, compatible with multi-position vacuum manifolds, and comply with ISO 9001 standards.
  • Features packing material held by two polyethylene frits.
  • Designed with a female Luer inlet that accepts a male Luer tip.
  • Molded for precise dimensions, suitable for automation equipment.
  • Sorbent chemistries tested for chromatographic activities, retention, and selectivity.