Learn More
Waters Corp Sep-Pak Cartridge, tC2, Plus Light, 145 mg Sorbent, 37–55 μm
Catalog No. 50785663 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This solid phase extraction cartridge features a silica-based bonded phase with low hydrophobicity, suitable for methods requiring less retention than C8 and applications similar to C18 and C8. It is ideal for various samples in GC, HPLC, and UPLC analysis.
  • Suitable for various samples in GC, HPLC, and UPLC analysis.
  • Offers a silica-based bonded phase with low hydrophobicity.
  • Functions effectively for procedures needing less retention than C8, comparable to C18 and C8.
  • Provides consistent quality for liquid chromatography.
  • Manufactured under stringent cGMP, ISO 9001, and ISO 3485 requirements.
  • Ensures consistent batch-to-batch performance via quality control testing with protein standards.
  • Utilizes chemically inert materials to prevent contaminants and ensure interference-free readings.
Catalog No. 50-785-663 Supplier Waters Corp Supplier No. WAT052725
This solid phase extraction cartridge features a silica-based bonded phase with low hydrophobicity, suitable for methods requiring less retention than C8 and applications similar to C18 and C8. It is ideal for various samples in GC, HPLC, and UPLC analysis.
  • Suitable for various samples in GC, HPLC, and UPLC analysis.
  • Offers a silica-based bonded phase with low hydrophobicity.
  • Functions effectively for procedures needing less retention than C8, comparable to C18 and C8.
  • Provides consistent quality for liquid chromatography.
  • Manufactured under stringent cGMP, ISO 9001, and ISO 3485 requirements.
  • Ensures consistent batch-to-batch performance via quality control testing with protein standards.
  • Utilizes chemically inert materials to prevent contaminants and ensure interference-free readings.