Learn More
Waters Corp Waters Vac Cartridge, Aminopropyl (NH2), 6 cc, 1 g Sorbent, 55–105 μm
Catalog No. 50179650 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Sep-Pak Aminopropyl (NH2) 6 cc Vac Cartridge features a silica-based polar bonded phase with basic character. It functions as a polar sorbent with different selectivity for acidic/basic analytes or as a weak anion exchanger in aqueous media.
  • Silica-based polar bonded phase with basic character.
  • Can be used as a polar sorbent or weak anion exchanger.
  • Applications include phenols, phenolic pigments, petroleum fractionation, saccharides, drugs, and metabolites.
  • Each cartridge contains 1 g of sorbent.
  • Optimal particle size distribution, pore size, and surface area.
  • Packing is clean, dry, and reproducible for uniform results.
  • Works best in the 2 to 8 pH range.
  • Ships with 300 cartridges per package.
Catalog No. 50-179-650 Supplier Waters Corp Supplier No. 186006327
The Sep-Pak Aminopropyl (NH2) 6 cc Vac Cartridge features a silica-based polar bonded phase with basic character. It functions as a polar sorbent with different selectivity for acidic/basic analytes or as a weak anion exchanger in aqueous media.
  • Silica-based polar bonded phase with basic character.
  • Can be used as a polar sorbent or weak anion exchanger.
  • Applications include phenols, phenolic pigments, petroleum fractionation, saccharides, drugs, and metabolites.
  • Each cartridge contains 1 g of sorbent.
  • Optimal particle size distribution, pore size, and surface area.
  • Packing is clean, dry, and reproducible for uniform results.
  • Works best in the 2 to 8 pH range.
  • Ships with 300 cartridges per package.