Learn More
Waters Corp Waters Corporation Sep-Pak Cartridge, Aminopropyl (NH2), 500 mg, 6 cc
Catalog No. 50179649 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This chromatographic cartridge is designed for sample enrichment and purification. It features a silica-based polar bonded phase with basic character, allowing it to function as a polar sorbent or a weak anion exchanger. It ensures convenient transfer of samples and solutions, preventing spillage or evaporation. High-quality packing materials offer optimal surface area, pore size, and particle size distributions.
  • Silica-based polar bonded phase with basic character for varied selectivity.
  • Functions as a polar sorbent or a weak anion exchanger in aqueous mediums.
  • Applicable for phenols, petroleum fractionation, saccharides, drugs and metabolites.
  • Enables convenient transfer of samples and solutions without spillage or evaporation.
  • High-quality, hygienic, dry, and reusable packing materials.
  • Equipped with a 20 μm frit for good flow, low extractables, and depth filtration.
Catalog No. 50-179-649 Supplier Waters Corp Supplier No. 186006326
This chromatographic cartridge is designed for sample enrichment and purification. It features a silica-based polar bonded phase with basic character, allowing it to function as a polar sorbent or a weak anion exchanger. It ensures convenient transfer of samples and solutions, preventing spillage or evaporation. High-quality packing materials offer optimal surface area, pore size, and particle size distributions.
  • Silica-based polar bonded phase with basic character for varied selectivity.
  • Functions as a polar sorbent or a weak anion exchanger in aqueous mediums.
  • Applicable for phenols, petroleum fractionation, saccharides, drugs and metabolites.
  • Enables convenient transfer of samples and solutions without spillage or evaporation.
  • High-quality, hygienic, dry, and reusable packing materials.
  • Equipped with a 20 μm frit for good flow, low extractables, and depth filtration.