Learn More
Waters Corp Sep-Pak AC2 Plus Short Cartridge, 400 mg Sorbent, 85 μm, 50/pk
Catalog No. 50783716 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
AC2 Plus Short cartridges provide reliable solid phase extraction with low ash activated carbon. They concentrate pesticides and herbicides in water/food samples, enabling efficient processing.
  • Low ash activated carbon.
  • Concentrates pesticides and herbicides in water/food.
  • Used in Japan Pesticide Method #17.
  • Short body for easy pump/syringe attachment, positive-pressure flow.
  • Compatible with vacuum-assisted devices and automated systems.
  • Highly hydrophobic.
  • Optimized for 85 μm particle size analytes.
  • Removes/enriches polar organic molecules from water.
  • Reduces solvent waste and background interference.
  • Seamless integration into sample prep protocols.
  • Manufactured to cGMP, ISO 9001, and ISO 13485 standards.
  • Thoroughly QC tested.
Catalog No. 50-783-716 Supplier Waters Corp Supplier No. JJAN20229
AC2 Plus Short cartridges provide reliable solid phase extraction with low ash activated carbon. They concentrate pesticides and herbicides in water/food samples, enabling efficient processing.
  • Low ash activated carbon.
  • Concentrates pesticides and herbicides in water/food.
  • Used in Japan Pesticide Method #17.
  • Short body for easy pump/syringe attachment, positive-pressure flow.
  • Compatible with vacuum-assisted devices and automated systems.
  • Highly hydrophobic.
  • Optimized for 85 μm particle size analytes.
  • Removes/enriches polar organic molecules from water.
  • Reduces solvent waste and background interference.
  • Seamless integration into sample prep protocols.
  • Manufactured to cGMP, ISO 9001, and ISO 13485 standards.
  • Thoroughly QC tested.