Learn More
Waters Corp Sep-Pak Accell Plus QMA Short Cartridge, 360 mg, 37–55 μm
Catalog No. 50785104 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These cartridges feature a silica-based, hydrophilic, strong anion-exchanger with a large pore size (300Å). Designed for extracting anionic analytes in both aqueous and non-aqueous solutions, they are ideal for isolating larger molecules like anionic proteins, including immunoglobulins and enzymes. They support various applications, including isolating phenolic compounds and removing acidic pigments from food extracts. The cartridges easily attach to pumps and syringes for efficient processing or can be adapted for vacuum-assisted and automated systems.
  • Each package contains 50 cartridges.
  • Sorbent weight is 360 mg per cartridge.
  • Particle size range is 37–55 μm.
  • Operating pH range is 2 to 8.
  • Hold up volume is 0.8 mL.
  • Ion exchange capacity is 230 μeq/gram.
Catalog No. 50-785-104 Supplier Waters Corp Supplier No. WAT020545
These cartridges feature a silica-based, hydrophilic, strong anion-exchanger with a large pore size (300Å). Designed for extracting anionic analytes in both aqueous and non-aqueous solutions, they are ideal for isolating larger molecules like anionic proteins, including immunoglobulins and enzymes. They support various applications, including isolating phenolic compounds and removing acidic pigments from food extracts. The cartridges easily attach to pumps and syringes for efficient processing or can be adapted for vacuum-assisted and automated systems.
  • Each package contains 50 cartridges.
  • Sorbent weight is 360 mg per cartridge.
  • Particle size range is 37–55 μm.
  • Operating pH range is 2 to 8.
  • Hold up volume is 0.8 mL.
  • Ion exchange capacity is 230 μeq/gram.