Learn More
Waters Corp Waters Sep-Pak Alumina B Plus Long Cartridge, 1710 mg Sorbent, 50 to 300 μm
Catalog No. 50785096 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Alumina B Plus Long cartridges offer a highly active alumina sorbent with basic surface chemistry for diverse extraction needs. They are stable under high pH conditions, act as low-capacity ion exchangers, and are unaffected by high-energy radioactivity. Supporting both positive-pressure and vacuum-assisted flow, they are sealed in poly-foil pouches.
  • Highly active alumina with basic surface chemistry for strong analyte retention.
  • Stable under high pH; low-capacity ion exchanger.
  • Unaffected by high-energy radioactivity, for isolating radioactive compounds.
  • Extracts phospholipids, steroids, catecholamines, pesticides, herbicides, and priority pollutants.
  • Supports positive-pressure (syringes/pumps) and vacuum-assisted flow.
  • Sealed in poly-foil pouch for product integrity and purity.
Catalog No. 50-785-096 Supplier Waters Corp Supplier No. WAT020505
Alumina B Plus Long cartridges offer a highly active alumina sorbent with basic surface chemistry for diverse extraction needs. They are stable under high pH conditions, act as low-capacity ion exchangers, and are unaffected by high-energy radioactivity. Supporting both positive-pressure and vacuum-assisted flow, they are sealed in poly-foil pouches.
  • Highly active alumina with basic surface chemistry for strong analyte retention.
  • Stable under high pH; low-capacity ion exchanger.
  • Unaffected by high-energy radioactivity, for isolating radioactive compounds.
  • Extracts phospholipids, steroids, catecholamines, pesticides, herbicides, and priority pollutants.
  • Supports positive-pressure (syringes/pumps) and vacuum-assisted flow.
  • Sealed in poly-foil pouch for product integrity and purity.