Learn More
Waters Corp Waters Sep-Pak C18 Plus Short Cartridge, 360 mg Sorbent, 55–105 μm
Catalog No. 50818645 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This solid phase extraction cartridge with 360 mg of C18 sorbent (55–105 μm particle size) is designed for sample preparation. It adsorbs analytes of even weak hydrophobicity from aqueous solutions, functioning similarly to reversed-phase HPLC columns.
  • Silica-based bonded phase with strong hydrophobicity.
  • Suitable for robotic and manual applications.
  • Allows flow direction reversal for trace enrichment.
  • Reduces costs by consuming less solvent and reagent.
  • Increases recoveries through minimum sample transfer.
  • Enhances safety by easing disposal and reducing toxic exposure.
  • Improves accuracy, preventing cross-contamination.
  • Eliminates emulsion issues with simplified sample handling.
  • Designed for retaining non-polar and moderately polar compounds.
  • Compatible with various solvents like methanol, acetonitrile, and aqueous solutions.
Catalog No. 50-818-645 Supplier Waters Corp Supplier No. WAT020515
This solid phase extraction cartridge with 360 mg of C18 sorbent (55–105 μm particle size) is designed for sample preparation. It adsorbs analytes of even weak hydrophobicity from aqueous solutions, functioning similarly to reversed-phase HPLC columns.
  • Silica-based bonded phase with strong hydrophobicity.
  • Suitable for robotic and manual applications.
  • Allows flow direction reversal for trace enrichment.
  • Reduces costs by consuming less solvent and reagent.
  • Increases recoveries through minimum sample transfer.
  • Enhances safety by easing disposal and reducing toxic exposure.
  • Improves accuracy, preventing cross-contamination.
  • Eliminates emulsion issues with simplified sample handling.
  • Designed for retaining non-polar and moderately polar compounds.
  • Compatible with various solvents like methanol, acetonitrile, and aqueous solutions.