Learn More
Waters Corp Waters Sep-Pak C18 Plus Short Cartridge, 360 mg, 55–105 μm
Catalog No. 50785387 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Sep-Pak C18 Plus Short Cartridge features a silica-based bonded phase with strong hydrophobicity, designed to adsorb analytes, even those with weak hydrophobicity, from aqueous solutions. It behaves similarly to reversed-phase HPLC columns and is suitable for liquid and gas samples.
  • Can reverse flow direction for trace enrichment applications.
  • Suitable for use with liquid and gas samples.
  • Lowers costs due to less solvent and reagent consumption.
  • Increases recoveries with minimum sample transfer.
  • Enhances safety through ease of disposal and reduced exposure to toxic agents.
  • Improves accuracy by preventing cross-contamination and eliminating emulsion problems.
  • Contains high-quality packings with optimal surface area, pore-size, and particle-size distributions, designed for reproducible activity and capacity in sample preparation.
Catalog No. 50-785-387 Supplier Waters Corp Supplier No. WAT036575
Add to Cart
Add to Cart
Request a Quote
This Sep-Pak C18 Plus Short Cartridge features a silica-based bonded phase with strong hydrophobicity, designed to adsorb analytes, even those with weak hydrophobicity, from aqueous solutions. It behaves similarly to reversed-phase HPLC columns and is suitable for liquid and gas samples.
  • Can reverse flow direction for trace enrichment applications.
  • Suitable for use with liquid and gas samples.
  • Lowers costs due to less solvent and reagent consumption.
  • Increases recoveries with minimum sample transfer.
  • Enhances safety through ease of disposal and reduced exposure to toxic agents.
  • Improves accuracy by preventing cross-contamination and eliminating emulsion problems.
  • Contains high-quality packings with optimal surface area, pore-size, and particle-size distributions, designed for reproducible activity and capacity in sample preparation.