Learn More
Waters Corp Waters Corporation Solid Phase Extraction Cartridge, C8, 400 mg Sorbent, 37–55 μm, 50 pk
Catalog No. 50818647 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Sep-Pak C8 Plus Short Cartridge features a silica-based bonded phase with moderate hydrophobicity. It is designed for methods requiring less retention than C18, making it suitable for applications such as the analysis of drugs and their metabolites in serum, plasma or urine, and peptides in serum or plasma.
  • Silica sorbent substrate
  • C8 chemistry
  • Endcapped design
  • Plus Short Cartridge format
  • Mass Spec compatibility
  • Reversed phase mode
  • Particle size of 37–55 μm
  • Pore size of 125 Å
  • Sorbent weight of 400 mg
  • Not water wettable
  • pH range of 2 to 8
  • Hold up volume of 0.8 mL
Catalog No. 50-818-647 Supplier Waters Corp Supplier No. WAT036775
This Sep-Pak C8 Plus Short Cartridge features a silica-based bonded phase with moderate hydrophobicity. It is designed for methods requiring less retention than C18, making it suitable for applications such as the analysis of drugs and their metabolites in serum, plasma or urine, and peptides in serum or plasma.
  • Silica sorbent substrate
  • C8 chemistry
  • Endcapped design
  • Plus Short Cartridge format
  • Mass Spec compatibility
  • Reversed phase mode
  • Particle size of 37–55 μm
  • Pore size of 125 Å
  • Sorbent weight of 400 mg
  • Not water wettable
  • pH range of 2 to 8
  • Hold up volume of 0.8 mL