Learn More
Waters Corp Sep-Pak Cyanopropyl (CN) Plus Short Cartridge, 360 mg Sorbent per Cartridge, 55 - 105 µm, 50/pk
Catalog No. 50785103 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Silica-based polar bonded phase; can be used as less polar alternative to silica in normal-phase applications or as less hydrophobic alternative to C18 or C8 in reversed-phase applications; typical applications include drug metabolites and pesticides.
Catalog No. 50-785-103 Supplier Waters Corp Supplier No. WAT020540
Silica-based polar bonded phase; can be used as less polar alternative to silica in normal-phase applications or as less hydrophobic alternative to C18 or C8 in reversed-phase applications; typical applications include drug metabolites and pesticides.