Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Sep-Pak Diol Plus Short Cartridge, 360 mg, 37–55 μm

Catalog No. 50785101 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This cartridge provides a silica-based polar bonded phase with neutral character, serving as an alternative to silica in normal-phase applications where acidic properties are undesirable. It is also effective as a very weakly interacting phase in aqueous applications.

  • Suitable for automation equipment due to precise molded dimensions.
  • Color-coded ring assists in sorbent identification and cartridge sealing.
  • Compatible with mass spectrometry.
  • Contains 360 mg of Diol sorbent.
  • Particle size: 37–55 μm.
  • Pore size: 300 Å.
  • pH range: 2 to 8.
  • Hold up volume: 0.8 mL.
  • Applications include isolating antibiotics from cosmetics and proteins or peptides by hydrophobic interaction chromatography.

Catalog No. 50-785-101 Supplier Waters Corp Supplier No. WAT020530
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This cartridge provides a silica-based polar bonded phase with neutral character, serving as an alternative to silica in normal-phase applications where acidic properties are undesirable. It is also effective as a very weakly interacting phase in aqueous applications.

  • Suitable for automation equipment due to precise molded dimensions.
  • Color-coded ring assists in sorbent identification and cartridge sealing.
  • Compatible with mass spectrometry.
  • Contains 360 mg of Diol sorbent.
  • Particle size: 37–55 μm.
  • Pore size: 300 Å.
  • pH range: 2 to 8.
  • Hold up volume: 0.8 mL.
  • Applications include isolating antibiotics from cosmetics and proteins or peptides by hydrophobic interaction chromatography.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.