Learn More
Waters Corp Waters Sep-Pak Plus Long Cartridge, C18, 820 mg Sorbent, 55-105 μm, 50/pk
Catalog No. 50785197 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Sep-Pak C18 Plus Long Cartridges are designed for superior Solid Phase Extraction (SPE), featuring a silica-based bonded phase with strong hydrophobicity. They effectively adsorb analytes, even those with weak hydrophobicity, from aqueous solutions and behave similarly to reversed-phase HPLC columns.
  • Streamlines purchasing with customized quotes.
  • Provides superior sample preparation due to its recognition as a leading SPE product family.
  • Adsorbs analytes of even weak hydrophobicity from aqueous solutions.
  • Easy integration into existing sample preparation protocols.
  • Reduces background interference.
  • Reduces solvent waste by eliminating unnecessary solvent pre-washing steps.
  • Manufactured in an ISO 13485 and ISO 9002 registered facility, compliant with cGMP guidelines for Class 1 Medical Devices.
  • Compatible with other SPE equipment.
Catalog No. 50-785-197 Supplier Waters Corp Supplier No. WAT023635
Sep-Pak C18 Plus Long Cartridges are designed for superior Solid Phase Extraction (SPE), featuring a silica-based bonded phase with strong hydrophobicity. They effectively adsorb analytes, even those with weak hydrophobicity, from aqueous solutions and behave similarly to reversed-phase HPLC columns.
  • Streamlines purchasing with customized quotes.
  • Provides superior sample preparation due to its recognition as a leading SPE product family.
  • Adsorbs analytes of even weak hydrophobicity from aqueous solutions.
  • Easy integration into existing sample preparation protocols.
  • Reduces background interference.
  • Reduces solvent waste by eliminating unnecessary solvent pre-washing steps.
  • Manufactured in an ISO 13485 and ISO 9002 registered facility, compliant with cGMP guidelines for Class 1 Medical Devices.
  • Compatible with other SPE equipment.