Learn More
Waters Corp Waters Sep-Pak Florisil Cartridge, 910 mg Sorbent, 50-200 μm, 50/pk
Catalog No. 50785100 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Sep-Pak Florisil Plus Long Cartridge is a precision chromatographic tool designed for sample enrichment and purification. It utilizes Florisil™, a highly active, weakly basic sorbent, effective for adsorbing low to moderate polarity species from non-aqueous solutions. This design enables easy extraction and transfer of samples, minimizing spillage and evaporation loss.
  • Features Florisil™, a blend of magnesium oxide and silica gel.
  • Effective for adsorbing low to moderate polarity species from non-aqueous solutions.
  • Sealed in a non-contaminating poly-foil pouch for purity.
  • High-quality packings ensure reproducible activity and capacity.
  • Enables spill-free extraction and transfer of samples.
Catalog No. 50-785-100 Supplier Waters Corp Supplier No. WAT020525
The Sep-Pak Florisil Plus Long Cartridge is a precision chromatographic tool designed for sample enrichment and purification. It utilizes Florisil™, a highly active, weakly basic sorbent, effective for adsorbing low to moderate polarity species from non-aqueous solutions. This design enables easy extraction and transfer of samples, minimizing spillage and evaporation loss.
  • Features Florisil™, a blend of magnesium oxide and silica gel.
  • Effective for adsorbing low to moderate polarity species from non-aqueous solutions.
  • Sealed in a non-contaminating poly-foil pouch for purity.
  • High-quality packings ensure reproducible activity and capacity.
  • Enables spill-free extraction and transfer of samples.