Learn More
Waters Corp Waters Solid Phase Extraction Cartridge, Florisil 910 mg, 50 to 200 μm, Normal-phase
Catalog No. 50785418 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This solid phase extraction cartridge features a polar, highly active, weakly basic sorbent, specifically designed for the adsorption of low to moderate polarity species from nonaqueous solutions.
  • Florisil sorbent
  • Sorbent weight of 910 mg
  • Particle size of 50 to 200 μm
  • Plus Long Cartridge format
  • Mass Spec compatibility
  • Normal-phase operation
  • Pore size of 60 Å
  • Water wettable
  • Hold up volume of 1.4 mL
Catalog No. 50-785-418 Supplier Waters Corp Supplier No. WAT036955
Add to Cart
Add to Cart
Request a Quote
This solid phase extraction cartridge features a polar, highly active, weakly basic sorbent, specifically designed for the adsorption of low to moderate polarity species from nonaqueous solutions.
  • Florisil sorbent
  • Sorbent weight of 910 mg
  • Particle size of 50 to 200 μm
  • Plus Long Cartridge format
  • Mass Spec compatibility
  • Normal-phase operation
  • Pore size of 60 Å
  • Water wettable
  • Hold up volume of 1.4 mL