Learn More
Waters Corp Sep-Pak Aminopropyl (NH2) Plus Short Cartridge, 360 mg Sorbent per Cartridge, 55 - 105 µm, 50/pk
Catalog No. 50785102 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Sep-Pak Amino Propyl (NH2) Plus Short cartridges contain a moderately polar, silica-based bonded phase with weakly basic surface. The sorbent can be used as a polar sorbent with different selectivity for acidic/basic analytes or as a weak anion exchanges in aqueous medium below pH 8. Applications include extraction of phenols and phenolic pigments, petroleum fractionation, saccharides and drugs and metabolites. These versatile cartridges can attach easily to pumps and syringes for reliable, positive-pressure flow to speed up the sample processing time. They can also be fitted with removable reservoirs and used on vacuum-assisted flow devices and certain automated systems.
Catalog No. 50-785-102 Supplier Waters Corp Supplier No. WAT020535
Sep-Pak Amino Propyl (NH2) Plus Short cartridges contain a moderately polar, silica-based bonded phase with weakly basic surface. The sorbent can be used as a polar sorbent with different selectivity for acidic/basic analytes or as a weak anion exchanges in aqueous medium below pH 8. Applications include extraction of phenols and phenolic pigments, petroleum fractionation, saccharides and drugs and metabolites. These versatile cartridges can attach easily to pumps and syringes for reliable, positive-pressure flow to speed up the sample processing time. They can also be fitted with removable reservoirs and used on vacuum-assisted flow devices and certain automated systems.