Learn More
Waters Corp Waters Sep-Pak Silica Plus Long Cartridge, 690 mg Sorbent, 55-105 μm Particle Size, Normal-phase
Catalog No. 50785099 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Sep-Pak Silica Plus Long Cartridge is a solid phase extraction (SPE) product for efficient sample preparation. It features a polar silica sorbent, ideal for adsorbing analytes from non-polar solvents. With 690 mg sorbent and a 55-105 μm particle size, it offers a balance of separation efficiency and ease of use for normal-phase SPE applications.
  • Enhanced flow reversal for trace enrichment.
  • Suitable for liquid and gas samples.
  • Cost-effective due to reduced solvent use and increased recoveries.
  • Improved safety and accuracy; prevents cross-contamination.
  • Stackable design for increased capacity.
  • Rapid sample preparation; suitable for robotic and manual use.
  • Versatile for drugs, peptides, and environmental samples.
Catalog No. 50-785-099 Supplier Waters Corp Supplier No. WAT020520
The Sep-Pak Silica Plus Long Cartridge is a solid phase extraction (SPE) product for efficient sample preparation. It features a polar silica sorbent, ideal for adsorbing analytes from non-polar solvents. With 690 mg sorbent and a 55-105 μm particle size, it offers a balance of separation efficiency and ease of use for normal-phase SPE applications.
  • Enhanced flow reversal for trace enrichment.
  • Suitable for liquid and gas samples.
  • Cost-effective due to reduced solvent use and increased recoveries.
  • Improved safety and accuracy; prevents cross-contamination.
  • Stackable design for increased capacity.
  • Rapid sample preparation; suitable for robotic and manual use.
  • Versatile for drugs, peptides, and environmental samples.