Learn More
Waters Corp Waters SPE Cartridge, Sep-Pak tC18 Plus Short, 400 mg, Reversed-Phase
Catalog No. 50818646 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This solid phase extraction (SPE) cartridge is designed for sample preparation, offering strong hydrophobicity and increased hydrolytic stability due to its trifunctional bonding chemistry. This lab equipment streamlines purchasing and simplifies protocols by reducing the number of steps and minimizing emulsion problems.
  • tC18 chemistry, silica-based bonded phase with strong hydrophobicity.
  • 400 mg sorbent.
  • Particle size: 37 to 55 μm; Pore size: 125 Å.
  • Plus Short cartridge format; 50 cartridges per pack.
  • Increased hydrolytic stability.
  • Mass spectrometry compatible; Reversed phase mode.
  • pH range: 2 to 8.
  • Suitable for analyte isolation from biological, environmental, and food samples.
  • Reduces sample handling, enhances safety, facilitates ease of disposal and direct field sampling.
Catalog No. 50-818-646 Supplier Waters Corp Supplier No. WAT036810
This solid phase extraction (SPE) cartridge is designed for sample preparation, offering strong hydrophobicity and increased hydrolytic stability due to its trifunctional bonding chemistry. This lab equipment streamlines purchasing and simplifies protocols by reducing the number of steps and minimizing emulsion problems.
  • tC18 chemistry, silica-based bonded phase with strong hydrophobicity.
  • 400 mg sorbent.
  • Particle size: 37 to 55 μm; Pore size: 125 Å.
  • Plus Short cartridge format; 50 cartridges per pack.
  • Increased hydrolytic stability.
  • Mass spectrometry compatible; Reversed phase mode.
  • pH range: 2 to 8.
  • Suitable for analyte isolation from biological, environmental, and food samples.
  • Reduces sample handling, enhances safety, facilitates ease of disposal and direct field sampling.