Learn More
Waters Corp Sep-Pak SEC Desalting Cartridge, 1 cc, 5K MWCO, Start-Up Kit
Catalog No. 502391117 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Sep-Pak SEC Desalting Cartridge 1cc Start-Up Kit is designed for efficient purification, desalting, and buffer exchange of proteins with a 5,000 molecular weight cut-off. This kit supports up to 96 samples through gravity-assisted flow in a 1cc syringe barrel configuration, compatible with both manual and automated laboratory workflows for clean-up prior to optical or MS detection.
  • Desalts and buffer exchanges up to 96 samples.
  • Syringe barrel configuration for gravity-assisted flow.
  • Cartridge volume of 1 cc with 5,000 molecular weight cut-off.
  • Compatible with manual and automated workflows.
  • Ensures efficient clean-up for optical or mass spectrometry detection.
  • Quality control tested for ≥ 80% protein recovery.
Catalog No. 50-239-1117 Supplier Waters Corp Supplier No. 186010126
The Sep-Pak SEC Desalting Cartridge 1cc Start-Up Kit is designed for efficient purification, desalting, and buffer exchange of proteins with a 5,000 molecular weight cut-off. This kit supports up to 96 samples through gravity-assisted flow in a 1cc syringe barrel configuration, compatible with both manual and automated laboratory workflows for clean-up prior to optical or MS detection.
  • Desalts and buffer exchanges up to 96 samples.
  • Syringe barrel configuration for gravity-assisted flow.
  • Cartridge volume of 1 cc with 5,000 molecular weight cut-off.
  • Compatible with manual and automated workflows.
  • Ensures efficient clean-up for optical or mass spectrometry detection.
  • Quality control tested for ≥ 80% protein recovery.