Learn More
Waters Corp Sep-Pak SEC Desalting Cartridge, 1 cc, 5K MWCO, Classic Cartridge
Catalog No. 502391156 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This SEC desalting cartridge is designed for gravity-assisted flow, offering efficient purification, desalting, and buffer exchange for proteins with a molecular weight cut-off of 5,000. It can be used in both manual and automated workflows, providing efficient sample clean-up prior to optical or MS detection. Each cartridge is QC tested to ensure high protein recovery and reproducibility.
  • Designed for gravity-assisted flow and efficient sample clean-up.
  • Purifies, desalts, and performs buffer exchange for proteins with a molecular weight cut-off of 5,000.
  • Compatible with manual and automated workflows.
  • QC tested to ensure ≥ 80% protein recovery.
  • Packing material is a crosslinked spherical dextran-based resin, stable from pH 2–13.
  • Exclusion limit is ≥5 kDa for globular proteins and 20 base pairs for nucleic acids.
Catalog No. 50-239-1156 Supplier Waters Corp Supplier No. 186010127
This SEC desalting cartridge is designed for gravity-assisted flow, offering efficient purification, desalting, and buffer exchange for proteins with a molecular weight cut-off of 5,000. It can be used in both manual and automated workflows, providing efficient sample clean-up prior to optical or MS detection. Each cartridge is QC tested to ensure high protein recovery and reproducibility.
  • Designed for gravity-assisted flow and efficient sample clean-up.
  • Purifies, desalts, and performs buffer exchange for proteins with a molecular weight cut-off of 5,000.
  • Compatible with manual and automated workflows.
  • QC tested to ensure ≥ 80% protein recovery.
  • Packing material is a crosslinked spherical dextran-based resin, stable from pH 2–13.
  • Exclusion limit is ≥5 kDa for globular proteins and 20 base pairs for nucleic acids.