Learn More
Waters Corp Waters Sep-Pak tC18 Plate, 100 mg Sorbent, 96-well
Catalog No. 50781071 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Sep-Pak tC18 96-well plate features silica-based, strongly hydrophobic sorbent with trifunctional bonding chemistry, offering enhanced hydrolytic stability. Its design is compatible with most liquid handling and SPE automation systems, making it ideal for high-throughput laboratory applications.
  • High-purity polypropylene construction with quality sorbent-filled wells
  • Trifunctional bonding chemistry ensures effective analyte adsorption from aqueous solutions
  • Fluid capacity above top frit: Approximately 2 mL
  • Key specifications: tC18 chemistry, Endcapped, Mass spec compatible, Reversed phase mode
  • Particle size: 37 to 55 μm; Pore size: 125 Å
  • Sorbent details: Silica substrate, not water wettable, pH range 2 to 8, 17% carbon load
  • Manufactured under ISO 9001, ISO 13485, and cGMP standards, with rigorous QC testing
Catalog No. 50-781-071 Supplier Waters Corp Supplier No. 186002321
This Sep-Pak tC18 96-well plate features silica-based, strongly hydrophobic sorbent with trifunctional bonding chemistry, offering enhanced hydrolytic stability. Its design is compatible with most liquid handling and SPE automation systems, making it ideal for high-throughput laboratory applications.
  • High-purity polypropylene construction with quality sorbent-filled wells
  • Trifunctional bonding chemistry ensures effective analyte adsorption from aqueous solutions
  • Fluid capacity above top frit: Approximately 2 mL
  • Key specifications: tC18 chemistry, Endcapped, Mass spec compatible, Reversed phase mode
  • Particle size: 37 to 55 μm; Pore size: 125 Å
  • Sorbent details: Silica substrate, not water wettable, pH range 2 to 8, 17% carbon load
  • Manufactured under ISO 9001, ISO 13485, and cGMP standards, with rigorous QC testing