Learn More
Waters Corp Waters Sep-Pak tC18 96-well Plate, 25 mg Sorbent, 37–55 μm
Catalog No. 50781069 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Sep-Pak tC18 96-well plate features a silica-based bonded phase with strong hydrophobicity and trifunctional bonding chemistry, providing increased hydrolytic stability. Its innovative design is compatible with most liquid handling and Solid Phase Extraction (SPE) automation systems, making it suitable for high-throughput laboratory applications. The plate is molded from high-purity polypropylene.
  • Silica-based bonded phase with strong hydrophobicity for effective separation.
  • Trifunctional bonding chemistry provides increased hydrolytic stability.
  • Innovative 96-well plate design ensures compatibility with most liquid handling and SPE automation systems.
  • Manufactured in an ISO 13485 and ISO 9002-registered facility, meeting cGMP guidelines for Class 1 Medical Devices.
  • Thoroughly tested for quality to ensure reliable and consistent performance.
Catalog No. 50-781-069 Supplier Waters Corp Supplier No. 186002319
This Sep-Pak tC18 96-well plate features a silica-based bonded phase with strong hydrophobicity and trifunctional bonding chemistry, providing increased hydrolytic stability. Its innovative design is compatible with most liquid handling and Solid Phase Extraction (SPE) automation systems, making it suitable for high-throughput laboratory applications. The plate is molded from high-purity polypropylene.
  • Silica-based bonded phase with strong hydrophobicity for effective separation.
  • Trifunctional bonding chemistry provides increased hydrolytic stability.
  • Innovative 96-well plate design ensures compatibility with most liquid handling and SPE automation systems.
  • Manufactured in an ISO 13485 and ISO 9002-registered facility, meeting cGMP guidelines for Class 1 Medical Devices.
  • Thoroughly tested for quality to ensure reliable and consistent performance.