Learn More
Waters Corp Sep-Pak Solid Phase Extraction Cartridge, tC18, Short, Double Luer-Lock
Catalog No. 502392076 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Sep-Pak tC18 Plus Short Double Luer-Lock Cartridge is designed for sample preparation, featuring a silica-based bonded phase with strong hydrophobicity. Its trifunctional bonding chemistry provides enhanced hydrolytic stability, making its applications similar to C18.
  • Silica-based bonded phase with strong hydrophobicity.
  • Trifunctional bonding chemistry provides increased hydrolytic stability.
  • Applications are similar to C18.
  • Includes double Luer-Lock adaptors.
Catalog No. 50-239-2076 Supplier Waters Corp Supplier No. WAT036810DL
The Sep-Pak tC18 Plus Short Double Luer-Lock Cartridge is designed for sample preparation, featuring a silica-based bonded phase with strong hydrophobicity. Its trifunctional bonding chemistry provides enhanced hydrolytic stability, making its applications similar to C18.
  • Silica-based bonded phase with strong hydrophobicity.
  • Trifunctional bonding chemistry provides increased hydrolytic stability.
  • Applications are similar to C18.
  • Includes double Luer-Lock adaptors.