Learn More
Waters Corp Cartridge, tC18, Plus Long, Double Luer-Lock, 900 mg
Catalog No. 502269426 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product includes double Luer-Lock adaptors. It features a silica-based bonded phase with strong hydrophobicity and trifunctional bonding chemistry, which increases its hydrolytic stability. Its applications are similar to those of C18 cartridges.
  • Fully compliant with ISO 80369-7 standard Luer-Lock adaptors, enabling easy connection with any other ISO standard tubings, adaptors, and devices.
  • Secure Luer-locking connection prevents dislodgement due to high pressure and spillage, avoiding hazardous material cleanup.
  • Allows for safely connecting multiple Plus cartridges in-series.
  • Provides Luer-Tip to Luer-Lock connection capability.
  • Features a silica-based bonded phase with strong hydrophobicity and trifunctional bonding chemistry.
  • Applications are similar to those of C18 cartridges.
This item is not returnable. View return policy
Catalog No. 50-226-9426 Supplier Waters Corp Supplier No. WAT036800DL
This product includes double Luer-Lock adaptors. It features a silica-based bonded phase with strong hydrophobicity and trifunctional bonding chemistry, which increases its hydrolytic stability. Its applications are similar to those of C18 cartridges.
  • Fully compliant with ISO 80369-7 standard Luer-Lock adaptors, enabling easy connection with any other ISO standard tubings, adaptors, and devices.
  • Secure Luer-locking connection prevents dislodgement due to high pressure and spillage, avoiding hazardous material cleanup.
  • Allows for safely connecting multiple Plus cartridges in-series.
  • Provides Luer-Tip to Luer-Lock connection capability.
  • Features a silica-based bonded phase with strong hydrophobicity and trifunctional bonding chemistry.
  • Applications are similar to those of C18 cartridges.