Learn More
Waters Corp Waters Sep-Pak Accell Plus CM Vac Cartridge, 3 cc, 500 mg Sorbent, 37–55 μm
Catalog No. 50785120 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Sep-Pak Accell Plus CM Vac cartridges utilize a silica-based, hydrophilic weak cation-exchanger with a 300Å pore size, ideal for extracting cationic analytes in various solutions. These syringe-barrel cartridges are designed for use with vacuum manifolds and automated SPE instruments, also suitable for isolating cationic proteins, steroids, pesticides, and herbicides.
  • Silica-based, hydrophilic weak cation-exchanger with 300Å pore size.
  • Extracts cationic analytes from aqueous and non-aqueous solutions.
  • Ideal for isolating cationic proteins.
  • Syringe-barrel design compatible with vacuum manifolds and automated SPE instruments.
  • Effective for extracting steroids, pesticides, and herbicides.
Catalog No. 50-785-120 Supplier Waters Corp Supplier No. WAT020855
Sep-Pak Accell Plus CM Vac cartridges utilize a silica-based, hydrophilic weak cation-exchanger with a 300Å pore size, ideal for extracting cationic analytes in various solutions. These syringe-barrel cartridges are designed for use with vacuum manifolds and automated SPE instruments, also suitable for isolating cationic proteins, steroids, pesticides, and herbicides.
  • Silica-based, hydrophilic weak cation-exchanger with 300Å pore size.
  • Extracts cationic analytes from aqueous and non-aqueous solutions.
  • Ideal for isolating cationic proteins.
  • Syringe-barrel design compatible with vacuum manifolds and automated SPE instruments.
  • Effective for extracting steroids, pesticides, and herbicides.