Learn More
Waters Corp Waters Vac Cartridge, Accell Plus QMA, 6 cc, 1 g Sorbent, 37 to 55 μm
Catalog No. 50785760 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These cartridges feature a silica-based, hydrophilic, strong anion-exchanger with a large pore size (300Å). Designed for extracting anionic analytes in aqueous and non-aqueous solutions, they are ideal for isolating larger molecules such as proteins and removing acidic pigments in food extracts. Syringe-barrel type, they are compatible with vacuum manifolds and automated SPE instruments.
  • Silica-based, hydrophilic, strong anion-exchanger
  • Vac cartridge format, 6 cc barrel size
  • 1 g sorbent, 37 to 55 μm particle size
  • 300 Å pore size, 230 μeq/gram ion exchange capacity
  • 2 to 8 pH range
  • Water wettable and mass spec compatible
  • Ion-exchange mode for solid phase extraction
  • 30 units per package
  • For isolating larger molecules and extracting anionic analytes
  • Use with vacuum manifolds and automated SPE instruments
Catalog No. 50-785-760 Supplier Waters Corp Supplier No. WAT054595
These cartridges feature a silica-based, hydrophilic, strong anion-exchanger with a large pore size (300Å). Designed for extracting anionic analytes in aqueous and non-aqueous solutions, they are ideal for isolating larger molecules such as proteins and removing acidic pigments in food extracts. Syringe-barrel type, they are compatible with vacuum manifolds and automated SPE instruments.
  • Silica-based, hydrophilic, strong anion-exchanger
  • Vac cartridge format, 6 cc barrel size
  • 1 g sorbent, 37 to 55 μm particle size
  • 300 Å pore size, 230 μeq/gram ion exchange capacity
  • 2 to 8 pH range
  • Water wettable and mass spec compatible
  • Ion-exchange mode for solid phase extraction
  • 30 units per package
  • For isolating larger molecules and extracting anionic analytes
  • Use with vacuum manifolds and automated SPE instruments