Learn More
Waters Corp Waters Sep-Pak Accell Plus QMA Vac Cartridge, 3 mL, 37 to 55 μm
Catalog No. 50785119 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Sep-Pak Accell Plus QMA Vac cartridges are silica-based, hydrophilic, strong anion-exchangers designed for the extraction of anionic analytes from aqueous and non-aqueous solutions. Their large pore size (300Å) allows isolation of larger molecules, and they are suitable for use with vacuum manifolds and automated SPE instruments.
  • Silica-based, hydrophilic, strong anion-exchanger with large pore size (300Å).
  • Designed for extraction of anionic analytes in aqueous and non-aqueous solutions.
  • Large pore size is ideal for isolating larger molecules such as anionic proteins (immunoglobulins and enzymes).
  • Applications include isolating phenolic compounds and removing acidic pigments in wine, fruit juices, and food extracts.
  • Syringe-barrel-type cartridges designed for use with vacuum manifolds and automated SPE instruments.
Catalog No. 50-785-119 Supplier Waters Corp Supplier No. WAT020850
Sep-Pak Accell Plus QMA Vac cartridges are silica-based, hydrophilic, strong anion-exchangers designed for the extraction of anionic analytes from aqueous and non-aqueous solutions. Their large pore size (300Å) allows isolation of larger molecules, and they are suitable for use with vacuum manifolds and automated SPE instruments.
  • Silica-based, hydrophilic, strong anion-exchanger with large pore size (300Å).
  • Designed for extraction of anionic analytes in aqueous and non-aqueous solutions.
  • Large pore size is ideal for isolating larger molecules such as anionic proteins (immunoglobulins and enzymes).
  • Applications include isolating phenolic compounds and removing acidic pigments in wine, fruit juices, and food extracts.
  • Syringe-barrel-type cartridges designed for use with vacuum manifolds and automated SPE instruments.