Learn More
Waters Corp Sep-Pak Accell Plus QMA 12 cc Vac Cartridge, 2 g Sorbent per Cartridge, 37 - 55 µm, 20/pk
Catalog No. 50785769 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Sep-Pak Accell Plus QMA Vac cartridges contain a silica-based, hydrophilic, strong anion-exchanger with large pore size (300Å). The cartridges are designed for extraction of anionic analytes in aqueous and non-aqueous solutions. The large pore size makes it ideal for isolation of larger molecules such as anionic proteins such as immunoglobulins and enzymes. Other applications include isolation of phenolic compounds and removal of acidic pigments in wine, fruit juices, and food extracts. These syringe-barrel-type cartridges are designed for use with vacuum manifolds and automated SPE instruments.
Catalog No. 50-785-769 Supplier Waters Corp Supplier No. WAT054640
Sep-Pak Accell Plus QMA Vac cartridges contain a silica-based, hydrophilic, strong anion-exchanger with large pore size (300Å). The cartridges are designed for extraction of anionic analytes in aqueous and non-aqueous solutions. The large pore size makes it ideal for isolation of larger molecules such as anionic proteins such as immunoglobulins and enzymes. Other applications include isolation of phenolic compounds and removal of acidic pigments in wine, fruit juices, and food extracts. These syringe-barrel-type cartridges are designed for use with vacuum manifolds and automated SPE instruments.