Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Vac Cartridge, Accell Plus QMA, 6 cc, 500 mg Sorbent, 37 to 55 μm

Catalog No. 50785751 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Sep-Pak Accell Plus QMA Vac cartridges are silica-based, strong anion-exchangers for extracting anionic analytes from aqueous and non-aqueous solutions. They isolate large molecules like proteins and enzymes, and remove acidic pigments. These cartridges work with vacuum manifolds and automated SPE instruments, ensuring spill-free sample transfer and precise chromatographic purification.

  • Extracts anionic analytes; isolates large molecules (proteins, enzymes)
  • Syringe-barrel design compatible with vacuum manifolds and automated SPE
  • Facilitates spill-free sample transfer and chromatographic purification
  • Ion exchange capacity: 230 μeq/gram
  • Mass spec compatible
  • Particle size: 37 to 55 μm
  • Sorbent weight: 500 mg
  • pH range: 2 to 8
  • Barrel size: 6 cc
  • Hold up volume: 1.2 mL

Catalog No. 50-785-751 Supplier Waters Corp Supplier No. WAT054550
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sep-Pak Accell Plus QMA Vac cartridges are silica-based, strong anion-exchangers for extracting anionic analytes from aqueous and non-aqueous solutions. They isolate large molecules like proteins and enzymes, and remove acidic pigments. These cartridges work with vacuum manifolds and automated SPE instruments, ensuring spill-free sample transfer and precise chromatographic purification.

  • Extracts anionic analytes; isolates large molecules (proteins, enzymes)
  • Syringe-barrel design compatible with vacuum manifolds and automated SPE
  • Facilitates spill-free sample transfer and chromatographic purification
  • Ion exchange capacity: 230 μeq/gram
  • Mass spec compatible
  • Particle size: 37 to 55 μm
  • Sorbent weight: 500 mg
  • pH range: 2 to 8
  • Barrel size: 6 cc
  • Hold up volume: 1.2 mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.