Learn More
Waters Corp Waters Sep-Pak Vac Cartridge, Accell Plus QMA, 1 cc, 100 mg Sorbent, 37-55 μm
Catalog No. 50785195 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These Sep-Pak Accell Plus QMA Vac cartridges are silica-based, hydrophilic, strong anion-exchangers (300Å pore size). Designed for extraction of anionic analytes from aqueous/non-aqueous solutions and isolating large molecules like anionic proteins. Intended for use with vacuum manifolds and automated SPE instruments.
  • Silica-based, hydrophilic, strong anion-exchanger.
  • Extracts anionic analytes from aqueous and non-aqueous solutions.
  • Isolates large molecules like anionic proteins.
  • Syringe-barrel format for vacuum manifolds and automated SPE.
  • Contains 100 mg sorbent in a 1 cc cartridge, particle size 37 to 55 μm.
  • Vacuum format ensures fast, efficient sample processing.
  • Compatible with various sample matrices (biological, environmental).
  • Offers excellent retention and recovery for high-quality sample preparation.
Catalog No. 50-785-195 Supplier Waters Corp Supplier No. WAT023620
These Sep-Pak Accell Plus QMA Vac cartridges are silica-based, hydrophilic, strong anion-exchangers (300Å pore size). Designed for extraction of anionic analytes from aqueous/non-aqueous solutions and isolating large molecules like anionic proteins. Intended for use with vacuum manifolds and automated SPE instruments.
  • Silica-based, hydrophilic, strong anion-exchanger.
  • Extracts anionic analytes from aqueous and non-aqueous solutions.
  • Isolates large molecules like anionic proteins.
  • Syringe-barrel format for vacuum manifolds and automated SPE.
  • Contains 100 mg sorbent in a 1 cc cartridge, particle size 37 to 55 μm.
  • Vacuum format ensures fast, efficient sample processing.
  • Compatible with various sample matrices (biological, environmental).
  • Offers excellent retention and recovery for high-quality sample preparation.