Learn More
Waters Corp Sep-Pak Cartridge, Alumina A, 6 cc, 500 mg, 50 to 300 μm
Catalog No. 50785748 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Sep-Pak Alumina A 6 mL Vac Cartridge contains 500 mg of Alumina A sorbent with a particle size range of 50 to 300 μm. This cartridge is designed for various laboratory applications, particularly normal phase chromatography.
  • Contains Alumina A sorbent for normal phase chromatography
  • Cartridge volume of 6 mL
  • Sorbent weight of 500 mg
  • Particle size range of 50 to 300 μm
  • Pore size of 120 Å
  • Operates within a pH range of 0.0 to 10.0
  • Features fully porous particle morphology
  • Available in a pack quantity of 30 pieces
Catalog No. 50-785-748 Supplier Waters Corp Supplier No. WAT054535
The Sep-Pak Alumina A 6 mL Vac Cartridge contains 500 mg of Alumina A sorbent with a particle size range of 50 to 300 μm. This cartridge is designed for various laboratory applications, particularly normal phase chromatography.
  • Contains Alumina A sorbent for normal phase chromatography
  • Cartridge volume of 6 mL
  • Sorbent weight of 500 mg
  • Particle size range of 50 to 300 μm
  • Pore size of 120 Å
  • Operates within a pH range of 0.0 to 10.0
  • Features fully porous particle morphology
  • Available in a pack quantity of 30 pieces