Learn More
Waters Corp Sep-Pak Vac Cartridge, Alumina A, 6 cc, 1 g Sorbent
Catalog No. 50785757 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Alumina A 6 cc Vac Cartridge is designed for normal phase chromatography. It features a 1 g sorbent with a fully porous aluminum oxide matrix, making it suitable for various laboratory applications.
  • Designed for normal phase chromatography (NP)
  • Aluminum oxide matrix
  • Fully porous particle morphology
  • 120 Å pore size
  • pH range 0.0 to 10.0
  • 1 g sorbent per cartridge
  • 6 cc volume
  • Particle size 50 to 300 μm
  • Available in a packaging unit of 30 pieces
Catalog No. 50-785-757 Supplier Waters Corp Supplier No. WAT054580
This Alumina A 6 cc Vac Cartridge is designed for normal phase chromatography. It features a 1 g sorbent with a fully porous aluminum oxide matrix, making it suitable for various laboratory applications.
  • Designed for normal phase chromatography (NP)
  • Aluminum oxide matrix
  • Fully porous particle morphology
  • 120 Å pore size
  • pH range 0.0 to 10.0
  • 1 g sorbent per cartridge
  • 6 cc volume
  • Particle size 50 to 300 μm
  • Available in a packaging unit of 30 pieces