Learn More
Waters Corp Sep-Pak Alumina A 20 cc Vac Cartridge, 5 g Sorbent per Cartridge, 50 - 300 µm, 20/pk
Catalog No. 50785775 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Sep-Pak Alumina A Vac cartridges contain a highly active grade of alumina with an acidic surface chemistry. The aluminum oxide surface provides an extremely polar surface for analyte retention and has properties of a Lewis acid. This normal phase sorbent is similar in use to silica; however, it is slightly more stable than unfunctionalized silica under high pH conditions. This acidic alumina is also low-capacity ion exchanger in aqueous media. Unlike polymer-based ion exhangers, this sorbent is unaffected by high-energy radioactivity and can be used for radioactive compound isolation. These syringe-barrel-type cartridges are designed for use with vacuum manifolds and automated SPE instruments.
Catalog No. 50-785-775 Supplier Waters Corp Supplier No. WAT054670
Sep-Pak Alumina A Vac cartridges contain a highly active grade of alumina with an acidic surface chemistry. The aluminum oxide surface provides an extremely polar surface for analyte retention and has properties of a Lewis acid. This normal phase sorbent is similar in use to silica; however, it is slightly more stable than unfunctionalized silica under high pH conditions. This acidic alumina is also low-capacity ion exchanger in aqueous media. Unlike polymer-based ion exhangers, this sorbent is unaffected by high-energy radioactivity and can be used for radioactive compound isolation. These syringe-barrel-type cartridges are designed for use with vacuum manifolds and automated SPE instruments.