Learn More
Waters Corp Vac Cartridge, Alumina A 500 mg, 3 cc, 50 to 300 μm
Catalog No. 50785113 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters Sep-Pak Alumina A Vac cartridges are ideal for normal-phase separations and radioactive compound isolation. They feature a highly active, acidic alumina sorbent that provides an extremely polar surface for analyte retention. These cartridges also offer enhanced stability under high pH conditions and can function as low-capacity ion exchangers.
  • Highly active grade of alumina with acidic surface chemistry.
  • Aluminum oxide surface provides an extremely polar surface for analyte retention.
  • Acts as a Lewis acid for diverse applications.
  • More stable than unfunctionalized silica under high pH conditions.
  • Functions as a low-capacity ion exchanger in aqueous media.
  • Unaffected by high-energy radioactivity, suitable for radioactive compound isolation.
  • Syringe-barrel design for use with vacuum manifolds and automated SPE instruments.
Catalog No. 50-785-113 Supplier Waters Corp Supplier No. WAT020820
Waters Sep-Pak Alumina A Vac cartridges are ideal for normal-phase separations and radioactive compound isolation. They feature a highly active, acidic alumina sorbent that provides an extremely polar surface for analyte retention. These cartridges also offer enhanced stability under high pH conditions and can function as low-capacity ion exchangers.
  • Highly active grade of alumina with acidic surface chemistry.
  • Aluminum oxide surface provides an extremely polar surface for analyte retention.
  • Acts as a Lewis acid for diverse applications.
  • More stable than unfunctionalized silica under high pH conditions.
  • Functions as a low-capacity ion exchanger in aqueous media.
  • Unaffected by high-energy radioactivity, suitable for radioactive compound isolation.
  • Syringe-barrel design for use with vacuum manifolds and automated SPE instruments.