Learn More
Waters Corp Vac Cartridge, Alumina B, 100 mg, 1 cc, 50-300 μm
Catalog No. 50785187 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These Alumina B Vac cartridges feature a highly active alumina with a basic surface chemistry, providing a polar surface for analyte retention and behaving as a Lewis acid. They are designed for use with vacuum manifolds and automated solid phase extraction instruments.
  • Contains highly active grade of alumina with basic surface chemistry.
  • Provides extremely polar surface for analyte retention.
  • Behaves as a Lewis acid.
  • Offers greater stability than unfunctionalized silica under high pH conditions.
  • Functions as a low-capacity ion exchanger in aqueous media.
  • Extracts phospholipids, steroids, and catecholamines.
  • Used for isolating radioactive compounds as it is unaffected by high-energy radioactivity.
  • Designed for use with vacuum manifolds and automated solid phase extraction instruments.
Catalog No. 50-785-187 Supplier Waters Corp Supplier No. WAT023580
These Alumina B Vac cartridges feature a highly active alumina with a basic surface chemistry, providing a polar surface for analyte retention and behaving as a Lewis acid. They are designed for use with vacuum manifolds and automated solid phase extraction instruments.
  • Contains highly active grade of alumina with basic surface chemistry.
  • Provides extremely polar surface for analyte retention.
  • Behaves as a Lewis acid.
  • Offers greater stability than unfunctionalized silica under high pH conditions.
  • Functions as a low-capacity ion exchanger in aqueous media.
  • Extracts phospholipids, steroids, and catecholamines.
  • Used for isolating radioactive compounds as it is unaffected by high-energy radioactivity.
  • Designed for use with vacuum manifolds and automated solid phase extraction instruments.