Learn More
Waters Corp Waters Corporation Vac Cartridge, Alumina B, 5 g Sorbent, 20 cc, 50 to 300 μm
Catalog No. 50785774 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Vac cartridge features a highly active grade of alumina with basic surface chemistry, providing an extremely polar surface for analyte retention. It acts as a Lewis acid and a low–capacity ion exchanger in aqueous media. More stable than silica under high pH, it is designed for vacuum manifolds and automated SPE instruments.
  • Contains a highly active grade of alumina with basic surface chemistry.
  • Provides an extremely polar surface for analyte retention.
  • Possesses properties of a Lewis acid.
  • More stable than unfunctionalized silica under high pH conditions.
  • Functions as a low–capacity ion exchanger in aqueous media.
  • Can be used to extract phospholipids, steroids, and catecholamines.
  • Unaffected by high–energy radioactivity, allowing for radioactive compound isolation.
  • Designed for use with vacuum manifolds and automated SPE instruments.
Catalog No. 50-785-774 Supplier Waters Corp Supplier No. WAT054665
This Vac cartridge features a highly active grade of alumina with basic surface chemistry, providing an extremely polar surface for analyte retention. It acts as a Lewis acid and a low–capacity ion exchanger in aqueous media. More stable than silica under high pH, it is designed for vacuum manifolds and automated SPE instruments.
  • Contains a highly active grade of alumina with basic surface chemistry.
  • Provides an extremely polar surface for analyte retention.
  • Possesses properties of a Lewis acid.
  • More stable than unfunctionalized silica under high pH conditions.
  • Functions as a low–capacity ion exchanger in aqueous media.
  • Can be used to extract phospholipids, steroids, and catecholamines.
  • Unaffected by high–energy radioactivity, allowing for radioactive compound isolation.
  • Designed for use with vacuum manifolds and automated SPE instruments.