Learn More
Waters Corp Sep-Pak Alumina B 6 cc Vac Cartridge, 500 mg Sorbent per Cartridge, 50 - 300 µm, 30/pk
Catalog No. 50785749 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Sep-Pak Alumina B Vac cartridges contain a highly active grade of alumina with a basic surface chemistry. The aluminum oxide surface provides an extremely polar surface for analyte retention and has properties of a Lewis acid. This normal phase sorbent is similar in use to silica; however, it is slightly more stable than unfunctionalized silica under high pH conditions. This basic alumina is also low-capacity ion exchanger in aqueous media. Sep-Pak Alumina B Vac cartridges can be used to extract phospholipids, steroids, and catecholamines. Since Sep-Pak Alumina B is unaffected by high-energy radioactivity, these cartridges can be used for radioactive compound isolation. These syringe-barrel-type cartridges are designed for use with vacuum manifolds and automated SPE instruments.
Catalog No. 50-785-749 Supplier Waters Corp Supplier No. WAT054540
Sep-Pak Alumina B Vac cartridges contain a highly active grade of alumina with a basic surface chemistry. The aluminum oxide surface provides an extremely polar surface for analyte retention and has properties of a Lewis acid. This normal phase sorbent is similar in use to silica; however, it is slightly more stable than unfunctionalized silica under high pH conditions. This basic alumina is also low-capacity ion exchanger in aqueous media. Sep-Pak Alumina B Vac cartridges can be used to extract phospholipids, steroids, and catecholamines. Since Sep-Pak Alumina B is unaffected by high-energy radioactivity, these cartridges can be used for radioactive compound isolation. These syringe-barrel-type cartridges are designed for use with vacuum manifolds and automated SPE instruments.