Learn More
Waters Corp Waters Sep-Pak Vac Cartridge, Alumina B, 3 cc, 500 mg
Catalog No. 50785114 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Sep-Pak Alumina B Vac cartridges offer highly active alumina with basic surface chemistry for polar analyte retention. This normal phase sorbent is more stable than silica under high pH and acts as a low–capacity ion exchanger. Designed for vacuum manifolds and automated SPE instruments.
  • Highly active alumina with basic surface chemistry for polar analyte retention
  • More stable than silica under high pH; functions as low–capacity ion exchanger
  • Extracts phospholipids, steroids, and catecholamines
  • Unaffected by high–energy radioactivity, suitable for radioactive compound isolation
  • Solid Phase Extraction (SPE) vac cartridge, normal–phase mode
  • Sorbent weight: 500 mg; Barrel size: 3 cc
  • Particle size: 50 – 300 μm; pH range: 0 to 10
  • Mass spec compatible and water wettable
Catalog No. 50-785-114 Supplier Waters Corp Supplier No. WAT020825
Sep-Pak Alumina B Vac cartridges offer highly active alumina with basic surface chemistry for polar analyte retention. This normal phase sorbent is more stable than silica under high pH and acts as a low–capacity ion exchanger. Designed for vacuum manifolds and automated SPE instruments.
  • Highly active alumina with basic surface chemistry for polar analyte retention
  • More stable than silica under high pH; functions as low–capacity ion exchanger
  • Extracts phospholipids, steroids, and catecholamines
  • Unaffected by high–energy radioactivity, suitable for radioactive compound isolation
  • Solid Phase Extraction (SPE) vac cartridge, normal–phase mode
  • Sorbent weight: 500 mg; Barrel size: 3 cc
  • Particle size: 50 – 300 μm; pH range: 0 to 10
  • Mass spec compatible and water wettable