Learn More
Waters Corp Waters Corporation Sep-Pak Vac Cartridge, Alumina N, 5 g, 20 cc
Catalog No. 50785506 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Sep-Pak Alumina N Vac cartridges provide a highly active, neutral alumina sorbent with an extremely polar surface for analyte retention, acting as a Lewis acid. This normal phase sorbent offers greater stability than silica under high pH, making it ideal for applications like synthetic crude oil fractionation. These syringe-barrel cartridges are designed for efficient use with vacuum manifolds and automated Solid Phase Extraction (SPE) instruments.
  • Highly active alumina with neutral surface chemistry.
  • Extremely polar surface for analyte retention, acting as a Lewis acid.
  • Normal phase sorbent; more stable than silica under high pH.
  • Ideal for synthetic crude oil fractionation and other organic compound analyses.
  • Syringe-barrel design for vacuum manifolds and automated SPE instruments.
  • Quality controlled for repeatable separations and reduced background interferences.
Catalog No. 50-785-506 Supplier Waters Corp Supplier No. WAT043375
Sep-Pak Alumina N Vac cartridges provide a highly active, neutral alumina sorbent with an extremely polar surface for analyte retention, acting as a Lewis acid. This normal phase sorbent offers greater stability than silica under high pH, making it ideal for applications like synthetic crude oil fractionation. These syringe-barrel cartridges are designed for efficient use with vacuum manifolds and automated Solid Phase Extraction (SPE) instruments.
  • Highly active alumina with neutral surface chemistry.
  • Extremely polar surface for analyte retention, acting as a Lewis acid.
  • Normal phase sorbent; more stable than silica under high pH.
  • Ideal for synthetic crude oil fractionation and other organic compound analyses.
  • Syringe-barrel design for vacuum manifolds and automated SPE instruments.
  • Quality controlled for repeatable separations and reduced background interferences.