Learn More
Waters Corp Waters Sep-Pak Vac Cartridge, Alumina N, 500 mg, 3 mL
Catalog No. 50785115 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters Sep-Pak Alumina N 3 mL Vac Cartridges are normal phase solid phase extraction sorbents featuring an extremely polar aluminum oxide surface. They are designed for robust analyte retention and are stable under high pH conditions. Compatible with vacuum manifolds and automated SPE instruments, they suit diverse analytical applications like synthetic crude oil fractionation, food additive analysis, and pesticide studies.
  • Contains a highly active grade of alumina with a neutral surface chemistry.
  • Aluminum oxide surface provides an extremely polar surface for analyte retention.
  • Normal phase sorbent, stable under high pH conditions.
  • Designed for use with vacuum manifolds and automated SPE instruments.
  • Water wettable and mass spec compatible.
  • Certified cartridges offer repeatable separations and superior performance for ultra-low level analyte detection.
Catalog No. 50-785-115 Supplier Waters Corp Supplier No. WAT020830
Waters Sep-Pak Alumina N 3 mL Vac Cartridges are normal phase solid phase extraction sorbents featuring an extremely polar aluminum oxide surface. They are designed for robust analyte retention and are stable under high pH conditions. Compatible with vacuum manifolds and automated SPE instruments, they suit diverse analytical applications like synthetic crude oil fractionation, food additive analysis, and pesticide studies.
  • Contains a highly active grade of alumina with a neutral surface chemistry.
  • Aluminum oxide surface provides an extremely polar surface for analyte retention.
  • Normal phase sorbent, stable under high pH conditions.
  • Designed for use with vacuum manifolds and automated SPE instruments.
  • Water wettable and mass spec compatible.
  • Certified cartridges offer repeatable separations and superior performance for ultra-low level analyte detection.