Learn More
Waters Corp Waters Corporation Sep-Pak Alumina N Vac Cartridge, 100 mg Sorbent, 50 – 300 μm, 100/pk
Catalog No. 50785188 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Sep-Pak Alumina N Vac cartridges feature a highly active grade of alumina with neutral surface chemistry, offering an extremely polar surface for analyte retention. This normal phase sorbent is suitable for applications like synthetic crude oil fractionation and is designed for use with vacuum manifolds and automated SPE instruments.
  • Highly active alumina with neutral surface chemistry.
  • Extremely polar surface for analyte retention; possesses Lewis acid properties.
  • More stable than unfunctionalized silica under high pH conditions.
  • Suitable for synthetic crude oil fractionation.
  • Designed for vacuum manifolds and automated SPE instruments.
Catalog No. 50-785-188 Supplier Waters Corp Supplier No. WAT023585
Sep-Pak Alumina N Vac cartridges feature a highly active grade of alumina with neutral surface chemistry, offering an extremely polar surface for analyte retention. This normal phase sorbent is suitable for applications like synthetic crude oil fractionation and is designed for use with vacuum manifolds and automated SPE instruments.
  • Highly active alumina with neutral surface chemistry.
  • Extremely polar surface for analyte retention; possesses Lewis acid properties.
  • More stable than unfunctionalized silica under high pH conditions.
  • Suitable for synthetic crude oil fractionation.
  • Designed for vacuum manifolds and automated SPE instruments.