Learn More
Waters Corp SPE Cartridge, Alumina N, 6 mL, 500 mg
Catalog No. 50785747 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These Sep-Pak Alumina N Vac cartridges offer highly active alumina with neutral surface chemistry for polar analyte retention. They are more stable than unfunctionalized silica under high pH conditions and are designed for use with vacuum manifolds and automated SPE instruments.
  • Highly active grade of alumina with neutral surface chemistry
  • Extremely polar surface for analyte retention
  • Properties of a Lewis acid
  • More stable than unfunctionalized silica under high pH conditions
  • Suitable for applications such as synthetic crude oil fractionation
  • Syringe-barrel-type cartridges
  • Designed for use with vacuum manifolds and automated SPE instruments
Catalog No. 50-785-747 Supplier Waters Corp Supplier No. WAT054530
These Sep-Pak Alumina N Vac cartridges offer highly active alumina with neutral surface chemistry for polar analyte retention. They are more stable than unfunctionalized silica under high pH conditions and are designed for use with vacuum manifolds and automated SPE instruments.
  • Highly active grade of alumina with neutral surface chemistry
  • Extremely polar surface for analyte retention
  • Properties of a Lewis acid
  • More stable than unfunctionalized silica under high pH conditions
  • Suitable for applications such as synthetic crude oil fractionation
  • Syringe-barrel-type cartridges
  • Designed for use with vacuum manifolds and automated SPE instruments