Learn More
Waters Corp Waters Sep-Pak Vac Cartridge, Alumina N, 35 cc, 10 g Sorbent
Catalog No. 50785782 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Sep-Pak Alumina N Vac cartridges feature highly active alumina with neutral surface chemistry. The aluminum oxide surface offers extremely polar analyte retention and Lewis acid properties. This normal phase sorbent is more stable than unfunctionalized silica at high pH. Ideal for synthetic crude oil fractionation, these syringe-barrel cartridges work with vacuum manifolds and automated SPE instruments.
  • Highly active alumina with neutral surface chemistry.
  • Extremely polar aluminum oxide surface for analyte retention.
  • More stable than unfunctionalized silica at high pH.
  • Syringe-barrel cartridges for vacuum manifolds and automated SPE.
Catalog No. 50-785-782 Supplier Waters Corp Supplier No. WAT054705
Sep-Pak Alumina N Vac cartridges feature highly active alumina with neutral surface chemistry. The aluminum oxide surface offers extremely polar analyte retention and Lewis acid properties. This normal phase sorbent is more stable than unfunctionalized silica at high pH. Ideal for synthetic crude oil fractionation, these syringe-barrel cartridges work with vacuum manifolds and automated SPE instruments.
  • Highly active alumina with neutral surface chemistry.
  • Extremely polar aluminum oxide surface for analyte retention.
  • More stable than unfunctionalized silica at high pH.
  • Syringe-barrel cartridges for vacuum manifolds and automated SPE.