Learn More
Waters Corp Waters Sep-Pak Cartridge, Alumina N, 6 cc, 1 g, 50 to 300 μm
Catalog No. 50785756 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Sep-Pak Alumina N Vac cartridges feature active alumina with neutral surface chemistry. Their aluminum oxide surface provides polar analyte retention and exhibits Lewis acid properties, offering stability greater than unfunctionalized silica under high pH. These syringe-barrel cartridges are ideal for synthetic crude oil fractionation and use with vacuum manifolds and automated SPE instruments.
  • Facilitates easy sample extraction and transfer, minimizing spillage/loss.
  • Provides a dependable chromatographic tool for enrichment/purification.
  • Supports consistent positive-pressure flow with pumps/syringes.
  • Compatible with vacuum-assisted flow devices and automated systems.
  • Packaged in a non-contaminating poly-foil pouch for integrity and purity.
  • Constructed from inert polypropylene to prevent analyte reactions.
  • Operates efficiently across a wide pH range of 0 to 10.
Catalog No. 50-785-756 Supplier Waters Corp Supplier No. WAT054575
Sep-Pak Alumina N Vac cartridges feature active alumina with neutral surface chemistry. Their aluminum oxide surface provides polar analyte retention and exhibits Lewis acid properties, offering stability greater than unfunctionalized silica under high pH. These syringe-barrel cartridges are ideal for synthetic crude oil fractionation and use with vacuum manifolds and automated SPE instruments.
  • Facilitates easy sample extraction and transfer, minimizing spillage/loss.
  • Provides a dependable chromatographic tool for enrichment/purification.
  • Supports consistent positive-pressure flow with pumps/syringes.
  • Compatible with vacuum-assisted flow devices and automated systems.
  • Packaged in a non-contaminating poly-foil pouch for integrity and purity.
  • Constructed from inert polypropylene to prevent analyte reactions.
  • Operates efficiently across a wide pH range of 0 to 10.