Learn More
Waters Corp Waters Vac Cartridge, C18, 35 cc, 10 g Sorbent, 55 to 105 μm
Catalog No. 50818753 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This silica-based bonded phase effectively adsorbs both polar and nonpolar analytes, including those with weak hydrophobicity, from aqueous solutions. It is ideal for applications like drug metabolite analysis, peptide desalting, and trace organic analysis in water samples.
  • Silica-based bonded phase with strong hydrophobicity
  • Adsorbs analytes (even weakly hydrophobic) from aqueous solutions
  • Monofunctional bonding offers alternate selectivity
  • Manufactured to ISO 9002, ISO 13485, and cGMP standards
  • Convenient, reproducible, and disposable solid-phase extraction (SPE) device
  • Clean, dry, and reproducible packings
  • General-purpose SPE for polar and nonpolar analytes
  • Applications: drug metabolite analysis, peptide desalting, trace organic analysis
Catalog No. 50-818-753 Supplier Waters Corp Supplier No. WAT043345
This silica-based bonded phase effectively adsorbs both polar and nonpolar analytes, including those with weak hydrophobicity, from aqueous solutions. It is ideal for applications like drug metabolite analysis, peptide desalting, and trace organic analysis in water samples.
  • Silica-based bonded phase with strong hydrophobicity
  • Adsorbs analytes (even weakly hydrophobic) from aqueous solutions
  • Monofunctional bonding offers alternate selectivity
  • Manufactured to ISO 9002, ISO 13485, and cGMP standards
  • Convenient, reproducible, and disposable solid-phase extraction (SPE) device
  • Clean, dry, and reproducible packings
  • General-purpose SPE for polar and nonpolar analytes
  • Applications: drug metabolite analysis, peptide desalting, trace organic analysis