Learn More
Waters Corp Waters Sep-Pak Vac Cartridge, C18, 12 cc, 2 g, 55–105 μm
Catalog No. 50785409 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This cartridge is designed to adsorb analytes with even weak hydrophobicity from aqueous solutions, utilizing a silica-based bonded phase with strong hydrophobicity. It functions similarly to reversed-phase HPLC columns.
  • Enables rapid sample preparation for various applications.
  • Suitable for both robotic applications and convenient manual use.
  • Features a standard male-Luer tip for compatibility with multi-position vacuum manifolds.
  • Constructed with molded, high purity polypropylene syringe-barrel bodies and two polyethylene frits for packing material containment.
  • Reduces operational costs, increases recoveries, and enhances safety during sample processing.
  • Improves accuracy by eliminating cross-contamination and emulsion issues.
  • Compatible with liquid and gas samples, supporting trace enrichment applications with reverse flow direction.
Catalog No. 50-785-409 Supplier Waters Corp Supplier No. WAT036915
This cartridge is designed to adsorb analytes with even weak hydrophobicity from aqueous solutions, utilizing a silica-based bonded phase with strong hydrophobicity. It functions similarly to reversed-phase HPLC columns.
  • Enables rapid sample preparation for various applications.
  • Suitable for both robotic applications and convenient manual use.
  • Features a standard male-Luer tip for compatibility with multi-position vacuum manifolds.
  • Constructed with molded, high purity polypropylene syringe-barrel bodies and two polyethylene frits for packing material containment.
  • Reduces operational costs, increases recoveries, and enhances safety during sample processing.
  • Improves accuracy by eliminating cross-contamination and emulsion issues.
  • Compatible with liquid and gas samples, supporting trace enrichment applications with reverse flow direction.