Learn More
Waters Corp Sep-Pak C18 1 cc Vac Cartridge, 100 mg Sorbent per Cartridge, 55 - 105 µm, 100/pk
Catalog No. 50785189 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Sep-Pak C18 Vac cartridges contain a hydrophobic, reverse-phase, silica-based bonded phase that is used to adsorb analytes of even weak hydrophobicity from aqueous solutions. The Sep-Pak C18 sorbent is made using the most reproducible bonding technology available, utilizing monofunctional silane chemistry. This sorbent offers a unique selectivity and excellent recovery of anlytes, and is recommended as a general purpose SPE phase for both polar and non-polar analytes. Typical applications include extraction of drugs and their metabolites in serum, plasma or urine, desalting of peptides, isolation of trace organics in environmental water samples and organic acids in beverages. These syringe-barrel-type cartridges are designed for use with vacuum manifolds and automated SPE instruments.
Catalog No. 50-785-189 Supplier Waters Corp Supplier No. WAT023590
Sep-Pak C18 Vac cartridges contain a hydrophobic, reverse-phase, silica-based bonded phase that is used to adsorb analytes of even weak hydrophobicity from aqueous solutions. The Sep-Pak C18 sorbent is made using the most reproducible bonding technology available, utilizing monofunctional silane chemistry. This sorbent offers a unique selectivity and excellent recovery of anlytes, and is recommended as a general purpose SPE phase for both polar and non-polar analytes. Typical applications include extraction of drugs and their metabolites in serum, plasma or urine, desalting of peptides, isolation of trace organics in environmental water samples and organic acids in beverages. These syringe-barrel-type cartridges are designed for use with vacuum manifolds and automated SPE instruments.