Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Vac Cartridge, C18, 1 cc, 50 mg Sorbent, 55–105 μm

Catalog No. 50780187 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This vac cartridge employs a silica-based C18 bonded phase with strong hydrophobicity for adsorbing analytes from aqueous solutions. It streamlines LC sample preparation, is compatible with multi-position vacuum manifolds, and is suited for robotic applications. High-quality sorbents ensure proper sample preparation and prevent contamination.

  • Sorbent type: C18, silica
  • Sorbent mass: 50 mg
  • Cartridge volume: 1 cc
  • Particle size: 55–105 μm
  • Endcapped
  • pH range: 2 to 8 pH
  • Hold up volume: 0.13 mL
  • Applications: drugs, metabolites, peptides, organics in water, organic acids in beverages.

Catalog No. 50-780-187 Supplier Waters Corp Supplier No. 186000308
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This vac cartridge employs a silica-based C18 bonded phase with strong hydrophobicity for adsorbing analytes from aqueous solutions. It streamlines LC sample preparation, is compatible with multi-position vacuum manifolds, and is suited for robotic applications. High-quality sorbents ensure proper sample preparation and prevent contamination.

  • Sorbent type: C18, silica
  • Sorbent mass: 50 mg
  • Cartridge volume: 1 cc
  • Particle size: 55–105 μm
  • Endcapped
  • pH range: 2 to 8 pH
  • Hold up volume: 0.13 mL
  • Applications: drugs, metabolites, peptides, organics in water, organic acids in beverages.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.