Learn More
Waters Corp Waters Sep-Pak C18 Vac Cartridge, 1 cc, 50 mg, 55–105 μm
Catalog No. 50785824 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
  • Hydrophobic, reverse-phase, silica-based bonded phase.
  • Utilizes monofunctional silane chemistry for reproducible bonding.
  • Offers unique selectivity and excellent recovery of analytes.
  • Recommended as a general-purpose Solid Phase Extraction (SPE) phase for both polar and non-polar analytes.
  • Syringe-barrel-type cartridges designed for use with vacuum manifolds and automated SPE instruments.
  • Sealed in a non-contaminating poly-foil pouch to maintain product integrity, packing inactivity, and purity.
Catalog No. 50-785-824 Supplier Waters Corp Supplier No. WAT054955
  • Hydrophobic, reverse-phase, silica-based bonded phase.
  • Utilizes monofunctional silane chemistry for reproducible bonding.
  • Offers unique selectivity and excellent recovery of analytes.
  • Recommended as a general-purpose Solid Phase Extraction (SPE) phase for both polar and non-polar analytes.
  • Syringe-barrel-type cartridges designed for use with vacuum manifolds and automated SPE instruments.
  • Sealed in a non-contaminating poly-foil pouch to maintain product integrity, packing inactivity, and purity.