Learn More
Waters Corp Waters Sep-Pak C18 Vac Cartridge, 20 cc, 5 g Sorbent, 55–105 μm
Catalog No. 50785411 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This silica-based bonded phase cartridge features strong hydrophobicity and is designed to adsorb analytes of even weak hydrophobicity from aqueous solutions. It behaves similarly to reversed-phase HPLC columns and is suitable for various applications, including the analysis of drugs and their metabolites in serum, plasma, or urine, desalting of peptides, trace organics in environmental water samples, and organic acids in beverages. The state-of-the-art packing process and precision design make it suitable for robotic applications and convenient for manual use.
  • Silica-based bonded phase with strong hydrophobicity.
  • Adsorbs analytes of weak hydrophobicity from aqueous solutions.
  • Similar in behavior to reversed-phase HPLC columns.
  • Suitable for robotic applications and convenient for manual use.
  • Ideal for sample enrichment and purification.
Catalog No. 50-785-411 Supplier Waters Corp Supplier No. WAT036925
This silica-based bonded phase cartridge features strong hydrophobicity and is designed to adsorb analytes of even weak hydrophobicity from aqueous solutions. It behaves similarly to reversed-phase HPLC columns and is suitable for various applications, including the analysis of drugs and their metabolites in serum, plasma, or urine, desalting of peptides, trace organics in environmental water samples, and organic acids in beverages. The state-of-the-art packing process and precision design make it suitable for robotic applications and convenient for manual use.
  • Silica-based bonded phase with strong hydrophobicity.
  • Adsorbs analytes of weak hydrophobicity from aqueous solutions.
  • Similar in behavior to reversed-phase HPLC columns.
  • Suitable for robotic applications and convenient for manual use.
  • Ideal for sample enrichment and purification.